siRNA Details
Gene Symbol: ZNF692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136036.2 | NP_001129508.1 | 9, 10 | 1464 | ||
NM_001193328.1 | NP_001180257.1 | 8, 9 | 1016 | |||
NM_017865.3 | NP_060335.2 | 9, 10 | 1248 | |||
XM_011544220.1 | XP_011542522.1 | 9, 10 | 1505 | |||
XM_011544221.1 | XP_011542523.1 | 9, 10 | 1502 | |||
XM_011544222.1 | XP_011542524.1 | 9, 10 | 1120 | |||
XM_011544223.2 | XP_011542525.1 | 8, 9 | 1370 | |||
XM_011544227.1 | XP_011542529.1 | 9, 10 | 1416 | |||
XM_017001698.1 | XP_016857187.1 | 9, 10 | 1117 | |||
XM_017001699.1 | XP_016857188.1 | 8, 9 | 1427 | |||
XM_017001700.1 | XP_016857189.1 | 8, 9 | 1042 | |||
XM_017001701.1 | XP_016857190.1 | 9, 10 | 1232 | |||
XM_017001702.1 | XP_016857191.1 | 8, 9 | 1157 | |||
XM_017001703.1 | XP_016857192.1 | 9, 10 | 1435 | |||
XM_017001704.1 | XP_016857193.1 | 9, 10 | 1392 | |||
XM_017001705.1 | XP_016857194.1 | 9, 10 | 1295 | |||
GenBank mRNA | AB209706.1 | - | 5, 6 | 1302 | ||
AF256175.1 | - | 2, 3 | 565 | |||
AK000538.1 | - | 8, 9 | 985 | |||
AK001070.1 | - | 7, 8 | 664 | |||
AK225535.1 | - | 8, 9 | 985 | |||
AK298899.1 | - | 7, 8 | 1193 | |||
AK302187.1 | - | 9, 10 | 1398 | |||
AL133100.1 | - | 7, 8 | 1977 | |||
BC002948.1 | - | 9, 10 | 1204 | 3533691 |
