siRNA Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194460.2 | NP_919442.1 | 4 | 374 | ||
| XM_005259594.1 | XP_005259651.1 | 4 | 454 | |||
| GenBank mRNA | AK000559.1 | - | 4 | 325 | ||
| AK289616.1 | - | 4 | 374 | |||
| BC001442.2 | - | 4 | 347 | 3138961 | ||
| BC025374.1 | - | 4 | 301 | 4093350 | ||
| BM558139.1 | - | 445 | ||||
| BM810439.1 | - | 444 | ||||

RNF126
Genomic Map