siRNA Details
| Gene Symbol: DENND4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017925.6 | NP_060395.5 | 28 | 5286 | ||
| NR_073201.3 | 28 | 5152 | ||||
| XM_006716804.3 | XP_006716867.1 | 29 | 5401 | |||
| XM_006716805.3 | XP_006716868.1 | 29 | 5401 | |||
| XM_006716807.3 | XP_006716870.1 | 20 | 3926 | |||
| XM_017014917.1 | XP_016870406.1 | 28 | 5194 | |||
| XM_017014918.1 | XP_016870407.1 | 30 | 5601 | |||
| XM_017014919.1 | XP_016870408.1 | 30 | 5568 | |||
| XM_017014920.1 | XP_016870409.1 | 29 | 5425 | |||
| GenBank mRNA | AK000693.1 | - | 10 | 1489 | ||
| AK001046.1 | - | 5 | 867 | |||
| AK002020.1 | - | 3 | 451 | |||
| AK025705.1 | - | 8 | 2307 | |||
| AK125842.1 | - | 22 | 4148 | |||
| AK127734.1 | - | 17 | 3562 | |||
| AK299842.1 | - | 15 | 3207 | |||
| AK302901.1 | - | 10 | 2618 | |||
| AL834521.1 | - | 7 | 1561 | |||
| BC144484.1 | - | 25 | 5008 | |||
| BC144485.1 | - | 24 | 4384 | |||
| CR627367.1 | - | 9 | 2492 | |||

DENND4C
Genomic Map