siRNA Details
| Gene Symbol: MFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033540.2 | NP_284941.2 | 14 | 1631 | ||
| XM_005247596.3 | XP_005247653.2 | 14 | 1589 | |||
| XM_011512963.2 | XP_011511265.1 | 11 | 1217 | |||
| GenBank mRNA | AB043588.1 | - | 13 | 1596 | ||
| AB043589.1 | - | 14 | 1596 | |||
| AF054986.1 | - | 4 | 388 | |||
| AF329637.1 | - | 13 | 1505 | |||
| AK000700.1 | - | 13 | 1461 | |||
| AK314306.1 | - | 14 | 1588 | |||
| BC040557.1 | - | 14 | 1631 | 5270347 | ||
| CR627379.1 | - | 11 | 1280 | |||

MFN1
Genomic Map