siRNA Details
| Gene Symbol: MFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033540.2 | NP_284941.2 | 2 | 182 | ||
| XM_005247596.3 | XP_005247653.2 | 2 | 140 | |||
| GenBank mRNA | AB043588.1 | - | 1 | 147 | ||
| AB043589.1 | - | 2 | 147 | |||
| AK314306.1 | - | 2 | 139 | |||
| BC020661.1 | - | 2 | 148 | |||
| BC040557.1 | - | 2 | 182 | 5270347 | ||

MFN1
Genomic Map