siRNA Details
| Gene Symbol: RADIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018059.4 | NP_060529.4 | 6 | 1732 | ||
| GenBank mRNA | AB058752.1 | - | 6 | 1676 | ||
| AK000997.1 | - | 2 | 170 | |||
| AK001186.1 | - | 3 | 715 | |||
| AK296325.1 | - | 5 | 1253 | |||
| AK302549.1 | - | 1724 | ||||
| BC117317.1 | - | 5 | 1618 | 40125868 | ||
| BC126311.1 | - | 5 | 1618 | 8992027 | ||
| BC143526.1 | - | 5 | 1618 | 9052035 | ||
| EF560723.1 | - | 5 | 1641 | |||

RADIL
Genomic Map