siRNA Details
| Gene Symbol: RADIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018059.4 | NP_060529.4 | 6, 7 | 1796 | ||
| GenBank mRNA | AB058752.1 | - | 6, 7 | 1740 | ||
| AK000997.1 | - | 2, 3 | 234 | |||
| AK001186.1 | - | 3, 4 | 779 | |||
| AK296325.1 | - | 5, 6 | 1317 | |||
| AK302549.1 | - | 1788 | ||||
| AL136731.1 | - | 1, 2 | 27 | |||
| BC117317.1 | - | 5, 6 | 1682 | 40125868 | ||
| BC126311.1 | - | 5, 6 | 1682 | 8992027 | ||
| BC143526.1 | - | 5, 6 | 1682 | 9052035 | ||
| EF560723.1 | - | 5, 6 | 1705 | |||

RADIL
Genomic Map