siRNA Details
| Gene Symbol: RADIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018059.4 | NP_060529.4 | 2 | 222 | ||
| GenBank mRNA | AB058752.1 | - | 2 | 172 | ||
| AK294745.1 | - | 2 | 182 | |||
| AK302549.1 | - | 153 | ||||
| BC117317.1 | - | 1 | 108 | 40125868 | ||
| BC126311.1 | - | 1 | 108 | 8992027 | ||
| BC143526.1 | - | 1 | 108 | 9052035 | ||
| BG722917.1 | - | 2 | 227 | |||
| CN365550.1 | - | 2 | 238 | |||
| DA241653.1 | - | 2 | 171 | |||
| EF560723.1 | - | 1 | 131 | |||

RADIL
Genomic Map