siRNA Details
Gene Symbol: IPO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018085.4 | NP_060555.2 | 23 | 3046 | ||
GenBank mRNA | AB033018.1 | - | 6 | 660 | ||
AF161391.1 | - | 6 | 699 | |||
AF410465.1 | - | 23 | 2996 | |||
AK001264.1 | - | 9 | 1218 | |||
AK027532.1 | - | 14 | 1910 | |||
AK074740.1 | - | 14 | 1927 | |||
AL834323.1 | - | 8 | 1103 | |||
BC003604.1 | - | 12 | 1782 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
