siRNA Details
| Gene Symbol: TSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018128.4 | NP_060598.3 | 5, 6 | 1943 | ||
| GenBank mRNA | AB037822.1 | - | 5, 6 | 2066 | ||
| AK001396.1 | - | 3, 4 | 585 | |||
| AK026565.1 | - | 5, 6 | 1016 | |||
| BC019090.2 | - | 4, 5 | 851 | |||
| BC110851.1 | - | 5, 6 | 1521 | 5786818 | ||
| BC126110.1 | - | 5, 6 | 1491 | 8991826 | ||

TSR1
Genomic Map