siRNA Details
| Gene Symbol: IQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160042.1 | NP_001153514.1 | 5 | 1549 | ||
| NM_018134.2 | NP_060604.2 | 5 | 1309 | |||
| GenBank mRNA | AK001409.1 | - | 5 | 1273 | ||
| AK223272.1 | - | 5 | 1297 | |||
| AK299235.1 | - | 4 | 1484 | |||
| AK307853.1 | - | 5 | 1513 | |||
| BC029986.1 | - | 1 | 437 | |||
| BC098250.1 | - | 6 | 763 | |||
| BC098260.1 | - | 5 | 1272 | 40024457 | ||
| BC098317.1 | - | 5 | 1272 | 40024460 | ||
| BC098366.1 | - | 5 | 1272 | 40024462 | ||

IQCC
Genomic Map