siRNA Details
| Gene Symbol: ASF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018154.2 | NP_060624.1 | 3 | 502 | ||
| GenBank mRNA | AB104486.1 | - | 3 | 330 | ||
| AF279307.1 | - | 3 | 423 | |||
| AK001288.1 | - | 3 | 461 | |||
| AK001466.1 | - | 3 | 493 | |||
| AK223080.1 | - | 3 | 456 | |||
| AK302134.1 | - | 3 | 463 | |||
| BC007726.2 | - | 3 | 426 | 4303318 | ||
| BC010014.2 | - | 3 | 436 | 4304756 | ||
| BC036521.1 | - | 3 | 466 | 5270216 | ||
| CR457235.1 | - | 3 | 330 | |||

ASF1B
Genomic Map