siRNA Details
| Gene Symbol: BTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002860.3 | NP_001002860.2 | 3 | 1099 | ||
| NM_018167.4 | NP_060637.1 | 3 | 1099 | |||
| XM_011536939.2 | XP_011535241.1 | 3 | 864 | |||
| GenBank mRNA | AB040958.2 | - | 3 | 1028 | ||
| AK001510.1 | - | 3 | 1025 | |||
| AK021953.1 | - | 1 | 318 | |||
| BC047071.1 | - | 3 | 1096 | |||
| BC141850.1 | - | 3 | 1040 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172806, AK033160, AK134271, BC057303, BC041669, and others.

BTBD7
Genomic Map