siRNA Details
| Gene Symbol: BTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002860.3 | NP_001002860.2 | 3 | 1268 | ||
| NM_018167.4 | NP_060637.1 | 3 | 1268 | |||
| XM_011536939.2 | XP_011535241.1 | 3 | 1033 | |||
| XM_017021438.1 | XP_016876927.1 | 1 | 33 | |||
| GenBank mRNA | AB040958.2 | - | 3 | 1197 | ||
| AK001510.1 | - | 3 | 1194 | |||
| AK021953.1 | - | 1 | 487 | |||
| AK303355.1 | - | 1 | 19 | |||
| BC047071.1 | - | 3 | 1265 | |||
| BC141850.1 | - | 3 | 1209 | |||

BTBD7
Genomic Map