siRNA Details
| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256944.1 | NP_001243873.1 | 9 | 1772 | ||
| NM_001830.3 | NP_001821.2 | 11 | 2027 | |||
| GenBank mRNA | AB019432.1 | - | 10 | 1872 | ||
| AF170492.1 | - | 9 | 1597 | |||
| AK225287.1 | - | 11 | 1988 | |||
| AK289564.1 | - | 11 | 1974 | |||
| AK299611.1 | - | 8 | 1405 | |||
| BC036068.1 | - | 10 | 1647 | 5311366 | ||
| BC130278.1 | - | 10 | 1886 | 40146309 | ||
| X77197.1 | - | 11 | 1980 | |||

CLCN4
Genomic Map