siRNA Details
| Gene Symbol: TMEM184C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018241.2 | NP_060711.2 | 5, 6 | 1138 | ||
| GenBank mRNA | AK000169.1 | - | 2, 3 | 291 | ||
| AK001708.1 | - | 5, 6 | 1081 | |||
| BC046128.1 | - | 5, 6 | 1140 | 5750613 | ||

TMEM184C
Genomic Map