siRNA Details
| Gene Symbol: TTC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307943.1 | NP_001294872.1 | 9 | 1147 | ||
| NM_018259.5 | NP_060729.2 | 9 | 1147 | |||
| XM_005253016.1 | XP_005253073.1 | 9 | 1147 | |||
| XM_006718263.1 | XP_006718326.1 | 7 | 830 | |||
| XM_011520218.2 | XP_011518520.1 | 9 | 1147 | |||
| XM_017018010.1 | XP_016873499.1 | 9 | 1147 | |||
| GenBank mRNA | AK001540.1 | - | 4 | 382 | ||
| AK001752.1 | - | 9 | 1100 | |||
| BC017238.2 | - | 9 | 1096 | 3940012 | ||
| BC033000.1 | - | 9 | 1093 | 4830377 | ||
| BX640972.1 | - | 4 | 987 | |||

TTC17
Genomic Map