siRNA Details
| Gene Symbol: TTC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307943.1 | NP_001294872.1 | 8, 9 | 1131 | ||
| NM_018259.5 | NP_060729.2 | 8, 9 | 1131 | |||
| XM_005253016.1 | XP_005253073.1 | 8, 9 | 1131 | |||
| XM_006718263.1 | XP_006718326.1 | 6, 7 | 814 | |||
| XM_011520218.2 | XP_011518520.1 | 8, 9 | 1131 | |||
| XM_017018010.1 | XP_016873499.1 | 8, 9 | 1131 | |||
| GenBank mRNA | AK001540.1 | - | 3, 4 | 366 | ||
| AK001752.1 | - | 8, 9 | 1084 | |||
| BC017238.2 | - | 8, 9 | 1080 | 3940012 | ||
| BC033000.1 | - | 8, 9 | 1077 | 4830377 | ||
| BX640972.1 | - | 3, 4 | 971 | |||

TTC17
Genomic Map