siRNA Details
| Gene Symbol: ZNF701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172655.1 | NP_001166126.1 | 5 | 1276 | ||
| NM_018260.2 | NP_060730.2 | 4 | 982 | |||
| XM_011527092.1 | XP_011525394.1 | 4 | 1056 | |||
| GenBank mRNA | AK001753.1 | - | 4 | 980 | ||
| BC080612.1 | - | 3 | 938 | 5816672 | ||
| BC131719.1 | - | 4 | 956 | 40123413 | ||
| BC136425.1 | - | 4 | 1086 | 9020414 | ||
| BC136426.1 | - | 4 | 954 | 9020415 | ||

ZNF701
Genomic Map