siRNA Details
| Gene Symbol: H2AFJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177925.3 | NP_808760.1 | 1 | 1371 | ||
| NR_027716.2 | 2 | 1057 | ||||
| GenBank mRNA | AK001765.1 | - | 2 | 817 | ||
| AK303301.1 | - | 3 | 934 | |||
| AL133626.1 | - | 2 | 811 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2AFJ| Gene Symbol: H2AFJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177925.3 | NP_808760.1 | 1 | 1371 | ||
| NR_027716.2 | 2 | 1057 | ||||
| GenBank mRNA | AK001765.1 | - | 2 | 817 | ||
| AK303301.1 | - | 3 | 934 | |||
| AL133626.1 | - | 2 | 811 | |||