siRNA Details
| Gene Symbol: PRR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018304.3 | NP_060774.2 | 9 | 1313 | ||
| GenBank mRNA | AK000296.1 | - | 6 | 629 | ||
| AK001891.1 | - | 9 | 1066 | |||
| AK225377.1 | - | 9 | 1066 | |||
| AK225380.1 | - | 9 | 1071 | |||
| BC008669.1 | - | 9 | 1055 | 3867293 | ||
| FJ161961.1 | - | 9 | 1066 | |||

PRR11
Genomic Map