siRNA Details
Gene Symbol: ERMARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278531.1 | NP_001265460.1 | 17 | 1669 | ||
NM_001278532.1 | NP_001265461.1 | 16 | 1641 | |||
NM_001278533.1 | NP_001265462.1 | 16 | 1591 | |||
NM_018341.2 | NP_060811.1 | 17 | 1810 | |||
XM_011535938.2 | XP_011534240.1 | 16 | 1780 | |||
XM_011535939.2 | XP_011534241.1 | 17 | 2900 | |||
XM_011535940.2 | XP_011534242.1 | 17 | 2709 | |||
XM_017011027.1 | XP_016866516.1 | 17 | 2650 | |||
XM_017011028.1 | XP_016866517.1 | 17 | 1975 | |||
XM_017011029.1 | XP_016866518.1 | 17 | 1735 | |||
XM_017011030.1 | XP_016866519.1 | 16 | 1559 | |||
XM_017011031.1 | XP_016866520.1 | 16 | 1624 | |||
XM_017011032.1 | XP_016866521.1 | 17 | 2895 | |||
XM_017011033.1 | XP_016866522.1 | 17 | 2704 | |||
GenBank mRNA | AK001369.1 | - | 16 | 1530 | ||
AK002014.1 | - | 17 | 1807 | |||
AK021520.1 | - | 7 | 1874 | |||
AK294090.1 | - | 16 | 1638 | |||
BC044812.1 | - | 16 | 1575 |
