siRNA Details
| Gene Symbol: CLCNKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042704.1 | NP_001036169.1 | 11, 12 | 1133 | ||
| NM_001257139.1 | NP_001244068.1 | 10, 11 | 1004 | |||
| NM_004070.3 | NP_004061.3 | 11, 12 | 1133 | |||
| GenBank mRNA | AK225550.1 | - | 11, 12 | 1088 | ||
| AK290814.1 | - | 11, 12 | 1088 | |||
| AK293476.1 | - | 1, 2 | 291 | |||
| AK293586.1 | - | 1, 2 | 334 | |||
| AK298285.1 | - | 10, 11 | 983 | |||
| BC035373.1 | - | 15, 16 | 1657 | |||
| BC048282.1 | - | 10, 11 | 1058 | 5184165 | ||
| BC053869.1 | - | 12, 13 | 1181 | 5187202 | ||
| Z30643.1 | - | 10, 11 | 1075 | |||
| Gene Symbol: CLCNKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000085.4 | NP_000076.2 | 11, 12 | 1205 | ||
| NM_001165945.2 | NP_001159417.2 | 4, 5 | 756 | |||
| GenBank mRNA | AK098217.1 | - | 4, 5 | 729 | ||
| AK292335.1 | - | 11, 12 | 1189 | |||
| BC020873.1 | - | 7, 8 | 780 | 4608625 | ||
| S80315.1 | - | 11, 12 | 1077 | |||
| Z30644.1 | - | 11, 12 | 1087 | |||

CLCNKA
Genomic Map