siRNA Details
| Gene Symbol: ST6GALNAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289107.1 | NP_001276036.1 | 10 | 2018 | ||
| NM_018414.4 | NP_060884.1 | 9 | 1881 | |||
| NR_110309.1 | 8 | 1777 | ||||
| GenBank mRNA | AK000113.1 | - | 8 | 1035 | ||
| AY096001.1 | - | 9 | 1681 | |||
| AY358918.1 | - | 9 | 1826 | |||
| BC022462.1 | - | 8 | 1632 | 4816713 | ||
| Y11339.2 | - | 9 | 1741 | |||

ST6GALNAC1
Genomic Map