siRNA Details
| Gene Symbol: ST6GALNAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289107.1 | NP_001276036.1 | 7 | 1711 | ||
| NM_018414.4 | NP_060884.1 | 6 | 1574 | |||
| XM_011524995.2 | XP_011523297.1 | 6 | 1494 | |||
| XM_011524996.2 | XP_011523298.1 | 6 | 1494 | |||
| XM_017024842.1 | XP_016880331.1 | 6 | 1494 | |||
| GenBank mRNA | AK000113.1 | - | 5 | 728 | ||
| AY096001.1 | - | 6 | 1374 | |||
| AY358918.1 | - | 6 | 1519 | |||
| Y11339.2 | - | 6 | 1434 | |||

ST6GALNAC1
Genomic Map