siRNA Details
| Gene Symbol: EMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018447.3 | NP_060917.1 | 9 | 1171 | ||
| GenBank mRNA | AF157321.1 | - | 8 | 883 | ||
| AK172843.1 | - | 8 | 879 | |||
| AK312008.1 | - | 8 | 878 | |||
| BC022807.1 | - | 8 | 866 | 4897577 | ||

EMC3
Genomic Map