siRNA Details
| Gene Symbol: EAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320041.1 | NP_001306970.1 | 3 | 387 | ||
| NM_018456.5 | NP_060926.2 | 4 | 524 | |||
| XM_005247618.3 | XP_005247675.1 | 5 | 674 | |||
| XM_017006861.1 | XP_016862350.1 | 5 | 396 | |||
| XM_017006862.1 | XP_016862351.1 | 3 | 296 | |||
| XM_017006863.1 | XP_016862352.1 | 4 | 467 | |||
| GenBank mRNA | AF217516.1 | - | 3 | 385 | ||
| AF517829.1 | - | 4 | 469 | |||
| AK301560.1 | - | 4 | 467 | |||
| AY049020.1 | - | 4 | 489 | |||
| BC014209.2 | - | 4 | 484 | 4765823 | ||

EAF2
Genomic Map