siRNA Details
| Gene Symbol: CISD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018464.4 | NP_060934.1 | 2 | 249 | ||
| XM_017016409.1 | XP_016871898.1 | 3 | 490 | |||
| GenBank mRNA | AF220049.1 | - | 2 | 188 | ||
| AK312017.1 | - | 2 | 124 | |||
| AY960578.1 | - | 2 | 132 | |||
| BC005962.1 | - | 2 | 225 | 4051044 | ||
| BC007043.1 | - | 2 | 181 | 3930701 | ||
| BC008474.1 | - | 2 | 186 | 4283253 | ||
| BC059168.1 | - | 2 | 196 | 4132410 | ||
| DA417986.1 | - | 2 | 249 | |||

CISD1
Genomic Map