siRNA Details
| Gene Symbol: CLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294338.1 | NP_001281267.1 | 4, 5 | 818 | ||
| NM_003993.3 | NP_003984.2 | 4, 5 | 815 | |||
| XM_005244878.1 | XP_005244935.1 | 4, 5 | 797 | |||
| XM_011509143.1 | XP_011507445.1 | 4, 5 | 800 | |||
| XM_011509144.1 | XP_011507446.1 | 4, 5 | 803 | |||
| XM_011509147.2 | XP_011507449.1 | 4, 5 | 790 | |||
| XM_017000241.1 | XP_016855730.1 | 4, 5 | 793 | |||
| XM_017000244.1 | XP_016855733.1 | 4, 5 | 787 | |||
| GenBank mRNA | AK291995.1 | - | 4, 5 | 662 | ||
| BC014067.2 | - | 4, 5 | 573 | 3837777 | ||
| BC053603.1 | - | 4, 5 | 736 | 6007375 | ||
| L29218.1 | - | 4, 5 | 618 | |||

CLK2
Genomic Map