siRNA Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278557.1 | NP_001265486.1 | 8 | 1057 | ||
| NM_001278558.1 | NP_001265487.1 | 6 | 780 | |||
| NM_018639.4 | NP_061109.1 | 8 | 1157 | |||
| XM_017019642.1 | XP_016875131.1 | 8 | 1059 | |||
| GenBank mRNA | AF038187.1 | - | 3 | 286 | ||
| AF163324.1 | - | 7 | 994 | |||
| AF229181.1 | - | 8 | 1096 | |||
| AK294230.1 | - | 6 | 814 | |||
| AK295555.1 | - | 8 | 1058 | |||
| AK298513.1 | - | 6 | 683 | |||
| AK310892.1 | - | 7 | 1155 | |||
| BC015887.2 | - | 8 | 1047 | 3910968 | ||

WSB2
Genomic Map