siRNA Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288590.1 | NP_001275519.1 | 3 | 902 | ||
| NM_001288591.1 | NP_001275520.1 | 3 | 902 | |||
| NM_001288592.1 | NP_001275521.1 | 2 | 361 | |||
| NM_018651.3 | NP_061121.2 | 3 | 939 | |||
| NM_025169.2 | NP_079445.1 | 3 | 939 | |||
| GenBank mRNA | AF154846.1 | - | 2 | 555 | ||
| AK022800.1 | - | 3 | 848 | |||
| AK291285.1 | - | 3 | 884 | |||
| AY280798.1 | - | 3 | 893 | |||
| BC010555.1 | - | 3 | 857 | |||
| BC063020.1 | - | 3 | 891 | |||
| BC120982.1 | - | 3 | 631 | |||
| BC120983.1 | - | 3 | 670 | |||
| BC127639.1 | - | 3 | 670 | |||
| BC127640.1 | - | 3 | 670 | |||
| BC148260.1 | - | 2 | 92 | |||

ZKSCAN7
Genomic Map