siRNA Details
| Gene Symbol: BIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018688.4 | NP_061158.1 | 9 | 735 | ||
| XM_005273573.3 | XP_005273630.1 | 8 | 641 | |||
| XM_011544586.2 | XP_011542888.1 | 10 | 820 | |||
| GenBank mRNA | AF271732.1 | - | 9 | 698 | ||
| AK001289.1 | - | 7 | 597 | |||
| AK023980.1 | - | 9 | 682 | |||
| AK124486.1 | - | 3 | 1020 | |||
| AK222470.1 | - | 9 | 707 | |||
| BC001223.1 | - | 8 | 619 | 3534031 | ||
| BC009824.2 | - | 9 | 669 | 3953604 | ||

BIN3
Genomic Map