siRNA Details
| Gene Symbol: BIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018688.4 | NP_061158.1 | 2, 3 | 155 | ||
| XM_005273573.3 | XP_005273630.1 | 2, 3 | 123 | |||
| XM_011544586.2 | XP_011542888.1 | 2, 3 | 127 | |||
| XM_011544587.1 | XP_011542889.1 | 2, 3 | 155 | |||
| GenBank mRNA | AF271732.1 | - | 2, 3 | 118 | ||
| AK023980.1 | - | 2, 3 | 102 | |||
| AK222470.1 | - | 2, 3 | 127 | |||
| BC001223.1 | - | 1, 2 | 39 | 3534031 | ||
| BC009824.2 | - | 2, 3 | 89 | 3953604 | ||

BIN3
Genomic Map