siRNA Details
| Gene Symbol: RCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136204.2 | NP_001129676.1 | 7 | 947 | ||
| NM_018715.3 | NP_061185.1 | 8 | 1116 | |||
| GenBank mRNA | AB040903.1 | - | 8 | 1047 | ||
| AJ421269.1 | - | 8 | 1133 | |||
| AL359612.1 | - | 5 | 466 | |||
| BC004933.1 | - | 4 | 387 | |||
| BC042141.1 | - | 7 | 912 | 5932549 | ||
| BC053908.1 | - | 7 | 945 | 6502476 | ||
| BC142946.1 | - | 7 | 703 | |||

RCC2
Genomic Map