siRNA Details
| Gene Symbol: FAM212B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019099.4 | NP_061972.1 | 2 | 495 | ||
| NM_198926.2 | NP_945120.1 | 2 | 353 | |||
| XM_011541783.2 | XP_011540085.1 | 4 | 591 | |||
| GenBank mRNA | AK055667.1 | - | 2 | 471 | ||
| AK293989.1 | - | 2 | 380 | |||
| AL137198.1 | - | 2 | 402 | |||
| AL137199.1 | - | 2 | 353 | |||
| BC031558.1 | - | 2 | 450 | 5181834 | ||
| DA306915.1 | - | 2 | 495 | |||

FAM212B
Genomic Map