siRNA Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019104.2 | NP_061977.1 | 8 | 983 | ||
| GenBank mRNA | AF162447.1 | - | 3 | 347 | ||
| AK293116.1 | - | 8 | 748 | |||
| BC009071.1 | - | 8 | 702 | 3872616 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIN37| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019104.2 | NP_061977.1 | 8 | 983 | ||
| GenBank mRNA | AF162447.1 | - | 3 | 347 | ||
| AK293116.1 | - | 8 | 748 | |||
| BC009071.1 | - | 8 | 702 | 3872616 | ||