siRNA Details
| Gene Symbol: SLC50A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122837.1 | NP_001116309.1 | 4 | 441 | ||
| NM_001122839.1 | NP_001116311.1 | 4 | 403 | |||
| NM_001287586.1 | NP_001274515.1 | 4 | 468 | |||
| NM_001287587.1 | NP_001274516.1 | 5 | 585 | |||
| NM_001287588.1 | NP_001274517.1 | 4 | 441 | |||
| NM_001287589.1 | NP_001274518.1 | 3 | 279 | |||
| NM_001287590.1 | NP_001274519.1 | 4 | 598 | |||
| NM_001287591.1 | NP_001274520.1 | 5 | 622 | |||
| NM_001287592.1 | NP_001274521.1 | 3 | 354 | |||
| NM_018845.3 | NP_061333.2 | 5 | 565 | |||
| XM_005245339.4 | XP_005245396.1 | 5 | 751 | |||
| XM_005245340.3 | XP_005245397.1 | 5 | 832 | |||
| XM_005245341.4 | XP_005245398.1 | 5 | 783 | |||
| XM_005245342.3 | XP_005245399.1 | 4 | 391 | |||
| XM_006711453.2 | XP_006711516.1 | 5 | 565 | |||
| GenBank mRNA | AF126023.1 | - | 5 | 525 | ||
| AF126024.1 | - | 4 | 390 | |||
| AK289551.1 | - | 5 | 566 | |||
| BC005943.1 | - | 5 | 509 | 4076256 | ||
| BC009621.1 | - | 4 | 365 | 3896154 | ||
| BC010869.1 | - | 4 | 356 | |||
| BC012378.1 | - | 4 | 390 | |||
| BE958344.1 | - | 422 | ||||
| BI195885.1 | - | 4 | 600 | |||
| BM806886.1 | - | 5 | 609 | |||
| BU160680.1 | - | 5 | 507 | |||
| BU564973.1 | - | 4 | 428 | |||
| BX093804.1 | - | 3 | 271 | |||
| CR457148.1 | - | 5 | 465 | |||
| X84712.1 | - | 3 | 150 | |||

SLC50A1
Genomic Map