siRNA Details
| Gene Symbol: NXF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032946.2 | NP_116564.2 | 10 | 1046 | ||
| NR_028089.1 | 10 | 1046 | ||||
| GenBank mRNA | AJ277654.1 | - | 10 | 1046 | ||
| AJ277655.1 | - | 10 | 1046 | |||
| AJ277656.1 | - | 10 | 982 | |||
| AJ277657.1 | - | 8 | 857 | |||
| AJ277658.1 | - | 8 | 857 | |||
| BC131708.1 | - | 9 | 867 | 40121743 | ||

NXF5
Genomic Map