siRNA Details
| Gene Symbol: SMG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005259057.2 | XP_005259114.1 | 8 | 1197 | ||
| XM_011527113.1 | XP_011525415.1 | 8 | 952 | |||
| XM_011527114.1 | XP_011525416.1 | 8 | 890 | |||
| XM_011527115.1 | XP_011525417.1 | 8 | 958 | |||
| XM_011527116.1 | XP_011525418.1 | 8 | 1084 | |||
| XM_017026988.1 | XP_016882477.1 | 4 | 308 | |||
| GenBank mRNA | AK022948.1 | - | 8 | 1169 | ||
| AK309830.1 | - | 8 | 1074 | |||
| AL136606.1 | - | 8 | 1123 | |||
| BC008869.2 | - | 8 | 1109 | 4131601 | ||

SMG9
Genomic Map