siRNA Details
| Gene Symbol: BARX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021570.3 | NP_067545.3 | 4 | 1192 | ||
| GenBank mRNA | AF213356.1 | - | 4 | 1182 | ||
| BC009458.1 | - | 3 | 1039 | |||
| BC064363.1 | - | 4 | 1237 | 5555949 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BARX1| Gene Symbol: BARX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021570.3 | NP_067545.3 | 4 | 1192 | ||
| GenBank mRNA | AF213356.1 | - | 4 | 1182 | ||
| BC009458.1 | - | 3 | 1039 | |||
| BC064363.1 | - | 4 | 1237 | 5555949 | ||