siRNA Details
| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019116.2 | NP_061989.2 | 5 | 867 | ||
| XM_011545894.2 | XP_011544196.1 | 5 | 873 | |||
| XM_017023459.1 | XP_016878948.1 | 5 | 873 | |||
| XM_017023460.1 | XP_016878949.1 | 5 | 873 | |||
| GenBank mRNA | AK092696.1 | - | 5 | 681 | ||
| AK124139.1 | - | 6 | 643 | |||

UBFD1
Genomic Map