siRNA Details
| Gene Symbol: PCDHB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018935.3 | NP_061758.1 | 1 | 1246 | ||
| GenBank mRNA | AF152494.1 | - | 1 | 1016 | ||
| AF217742.1 | - | 1 | 1016 | |||
| AK312794.1 | - | 1 | 1248 | |||
| BC038797.1 | - | 1 | 1182 | 5767743 | ||

PCDHB15
Genomic Map