siRNA Details
| Gene Symbol: PCDHB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018933.3 | NP_061756.1 | 1 | 1129 | ||
| GenBank mRNA | AF152492.1 | - | 1 | 924 | ||
| AF217745.1 | - | 1 | 924 | |||
| AK021915.1 | - | 1 | 1112 | |||
| AK292821.1 | - | 1 | 1091 | |||
| AK296151.1 | - | 1 | 1092 | |||
| AY358427.1 | - | 1 | 1094 | |||
| BC033068.1 | - | 1 | 1082 | 5446016 | ||
| BC051348.1 | - | 1 | 1082 | 6483735 | ||
| BC071934.1 | - | 1 | 1082 | 6383780 | ||

PCDHB13
Genomic Map