siRNA Details
| Gene Symbol: PCDHB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018930.3 | NP_061753.1 | 1 | 669 | ||
| GenBank mRNA | AF152489.1 | - | 1 | 495 | ||
| AF217748.1 | - | 1 | 495 | |||
| AK056391.1 | - | 1 | 679 | |||
| AK315583.1 | - | 1 | 695 | |||
| AY358720.1 | - | 1 | 673 | |||
| BC031837.1 | - | 1 | 669 | 4931995 | ||

PCDHB10
Genomic Map