siRNA Details
| Gene Symbol: PCDHA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018904.2 | NP_061727.1 | 1 | 882 | ||
| NM_031865.1 | NP_114071.1 | 1 | 882 | |||
| GenBank mRNA | AB208924.1 | - | 1 | 943 | ||
| AF152309.1 | - | 1 | 882 | |||
| AF152478.1 | - | 1 | 882 | |||

PCDHA13
Genomic Map