siRNA Details
| Gene Symbol: TEX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201457.1 | NP_001188386.1 | 14 | 2093 | ||
| NM_031272.4 | NP_112562.3 | 14 | 2075 | |||
| NM_198393.3 | NP_938207.2 | 14 | 2075 | |||
| XM_011525029.2 | XP_011523331.1 | 14 | 2054 | |||
| XM_017024860.1 | XP_016880349.1 | 12 | 1725 | |||
| XM_017024861.1 | XP_016880350.1 | 13 | 1900 | |||
| GenBank mRNA | AF285601.1 | - | 3 | 539 | ||
| AK125896.1 | - | 9 | 1041 | |||
| AK128217.1 | - | 10 | 1510 | |||
| AL834143.1 | - | 14 | 2074 | |||
| BC040526.1 | - | 14 | 2061 | 4831308 | ||

TEX14
Genomic Map