siRNA Details
| Gene Symbol: TEX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201457.1 | NP_001188386.1 | 18 | 3091 | ||
| NM_031272.4 | NP_112562.3 | 18 | 3073 | |||
| NM_198393.3 | NP_938207.2 | 18 | 3073 | |||
| XM_011525029.2 | XP_011523331.1 | 18 | 3175 | |||
| XM_017024860.1 | XP_016880349.1 | 16 | 2723 | |||
| XM_017024861.1 | XP_016880350.1 | 17 | 2898 | |||
| GenBank mRNA | AF285601.1 | - | 7 | 1537 | ||
| AK125896.1 | - | 13 | 2039 | |||
| AK128217.1 | - | 14 | 2508 | |||
| AK308299.1 | - | 18 | 3032 | |||
| AL834143.1 | - | 18 | 3072 | |||
| BC040526.1 | - | 18 | 3059 | 4831308 | ||

TEX14
Genomic Map