siRNA Details
| Gene Symbol: TEX13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291277.1 | NP_001278206.1 | 2 | 212 | ||
| NM_031274.4 | NP_112564.1 | 2 | 238 | |||
| GenBank mRNA | AF285597.1 | - | 2 | 219 | ||
| BC042547.1 | - | 2 | 193 | 5164310 | ||
| BC108734.1 | - | 2 | 211 | 5744723 | ||
| HY018872.1 | - | 2 | 212 | |||

TEX13A
Genomic Map