siRNA Details
| Gene Symbol: TEX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031275.4 | NP_112565.1 | 3 | 264 | ||
| GenBank mRNA | AF285600.1 | - | 2 | 170 | ||
| BC029506.1 | - | 3 | 264 | 5298531 | ||
| BI561514.1 | - | 3 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX12| Gene Symbol: TEX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031275.4 | NP_112565.1 | 3 | 264 | ||
| GenBank mRNA | AF285600.1 | - | 2 | 170 | ||
| BC029506.1 | - | 3 | 264 | 5298531 | ||
| BI561514.1 | - | 3 | 268 | |||