siRNA Details
| Gene Symbol: DNAH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018897.2 | NP_061720.2 | 17 | 2248 | ||
| XM_011511487.2 | XP_011509789.1 | 18 | 2624 | |||
| XM_011511488.2 | XP_011509790.1 | 16 | 3298 | |||
| XM_011511489.2 | XP_011509791.1 | 16 | 2210 | |||
| XM_011511490.2 | XP_011509792.1 | 17 | 3418 | |||
| XM_011511491.2 | XP_011509793.1 | 17 | 3420 | |||
| XM_011511492.2 | XP_011509794.1 | 17 | 3420 | |||
| XM_011511493.2 | XP_011509795.1 | 17 | 3420 | |||
| XM_011511494.2 | XP_011509796.1 | 17 | 3419 | |||
| XM_011511495.2 | XP_011509797.1 | 17 | 3419 | |||
| XM_017004504.1 | XP_016859993.1 | 16 | 3144 | |||
| GenBank mRNA | AB023161.2 | - | 17 | 2266 | ||
| AF327442.1 | - | 17 | 2243 | |||

DNAH7
Genomic Map